3-Chlor-4-hydroxybenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS40339G5EA 0 CHF
ACTIAS40339G5
3-Chlor-4-hydroxybenzaldehyd
3-Chloro-4-hydroxybenzaldehyde
Formel: C₇H₅ClO₂
MW: 156,57 g/mol
Siedepunkt: 251 °C (1013 hPa)
Melting Pt: 133…134 °C
Dichte: 1,0404 g/cm³ (20 °C)
CAS-Nummer: 2420-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR