6-Chlorosalicylaldehyde

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 2-Chloro-6-hydroxybenzaldehyde

ACTIAS30498G5EA 0 CHF
ACTIAS30498G5 ACTIAS30498G1
6-Chlorosalicylaldehyde
6-Chlorosalicylaldehyde
Formula: C₇H₅ClO₂
MW: 156.57 g/mol
MDL Number: MFCD01646166
CAS Number: 18362-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR