2-Chloro-4-hydroxybenzaldehyde

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS31023G5EA 0 CHF
ACTIAS31023G1 ACTIAS31023G5
2-Chloro-4-hydroxybenzaldehyde
2-Chloro-4-hydroxybenzaldehyde
Formula: C₇H₅ClO₂
MW: 156.57 g/mol
MDL Number: MFCD00052184
CAS Number: 56962-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR