2,5-Dibromo-3-nitropyridine

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS31183G5EA 0 CHF
ACTIAS31183G10 ACTIAS31183G5
2,5-Dibromo-3-nitropyridine
2,5-Dibromo-3-nitropyridine
Formula: Câ‚…Hâ‚‚Brâ‚‚Nâ‚‚Oâ‚‚
MDL Number: MFCD09266223
CAS Number: 15862-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR