2-Brom-5-nitro-4-picolin

Lieferant: ACTIVATE SCIENTIFIC GMBH

Synonyms: 2-Brom-4-methyl-5-nitropyridin

ACTIAS31573G5EA 0 CHF
ACTIAS31573G10 ACTIAS31573G5
2-Brom-5-nitro-4-picolin
2-Brom-5-nitro-4-picolin
Formel: C₆H₅BrN₂O₂
MW: 217,02 g/mol
MDL Number: MFCD03095066
CAS-Nummer: 23056-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR