2-Amino-4-bromophenol

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 5-Bromo-2-hydroxyaniline

ACTIAS31282G5EA 0 CHF
ACTIAS31282G5 ACTIAS31282G10
2-Amino-4-bromophenol
2-Amino-4-bromophenol
Formula: C₆H₆BrNO
MW: 188.02 g/mol
MDL Number: MFCD00235171
CAS Number: 40925-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR