3-Amino-4-bromopyridine

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 4-Bromo-3-pyridylamine4-Bromopyridin-3-amine

ACTIAS31215G1EA 0 CHF
ACTIAS31215G1 ACTIAS31215G5
3-Amino-4-bromopyridine
3-Amino-4-bromopyridine
Formula: Câ‚…Hâ‚…BrNâ‚‚
MDL Number: MFCD04112575
CAS Number: 239137-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR