4-Bromo-3-fluorobenzenesulphonamide

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS31902G5EA 0 CHF
ACTIAS31902G5 ACTIAS31902G1
4-Bromo-3-fluorobenzenesulphonamide
4-Bromo-3-fluorobenzenesulphonamide
Formula: C₆H₅BrFNO₂S
MW: 254.08 g/mol
MDL Number: MFCD03094292
CAS Number: 263349-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR