1-Bromo-2-fluoro-3-nitrobenzene

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 3-Bromo-2-fluoronitrobenzene

ACTIAS40315G5EA 0 CHF
ACTIAS40315G5
1-Bromo-2-fluoro-3-nitrobenzene
1-Bromo-2-fluoro-3-nitrobenzene
Formula: C₆H₃BrFNO₂
MW: 220 g/mol
MDL Number: MFCD05663754
CAS Number: 58534-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR