4-Bromo-5-fluoro-2-nitrobenzoic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS31126G5EA 0 CHF
ACTIAS31126G5 ACTIAS31126G25
4-Bromo-5-fluoro-2-nitrobenzoic acid
4-Bromo-5-fluoro-2-nitrobenzoic acid
Formula: C₇H₃BrFNO₄
MDL Number: MFCD08702773
CAS Number: 1020717-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR