4-Bromo-3-fluorobenzaldehyde

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS31159G5EA 0 CHF
ACTIAS31159G5 ACTIAS31159G1
4-Bromo-3-fluorobenzaldehyde
4-Bromo-3-fluorobenzaldehyde
Formula: C₇H₄BrFO
MW: 203.01 g/mol
MDL Number: MFCD03095000
CAS Number: 133059-43-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR