2-Bromo-5-fluoro-6-methylpyridine
Supplier: ACTIVATE SCIENTIFIC GMBH
Synonyms:
2-Bromo-5-fluoro-6-picoline
ACTIAS7197G5EA
0
CHF
ACTIAS7197G5
ACTIAS7197G25
2-Bromo-5-fluoro-6-methylpyridine
2-Bromo-5-fluoro-6-methylpyridine
Formula:
C₆H₅BrFN MW: 190.01 g/mol |
MDL Number:
MFCD03095064 CAS Number: 374633-38-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...