4-Bromo-3-nitrobenzoic acid
Supplier: ACTIVATE SCIENTIFIC GMBH
ACTIAS40409G5EA
0
CHF
ACTIAS40409G5
4-Bromo-3-nitrobenzoic acid
4-Bromo-3-nitrobenzoic acid
Formula:
C₇H₄BrNO₄ MW: 246.02 g/mol |
MDL Number:
MFCD00272137 CAS Number: 6319-40-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...