2-Bromo-4-chloro-1-fluorobenzene

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 1-Bromo-3-chloro-6-fluorobenzene2-Bromo-4-chlorofluorobenzene3-Bromo-1-chloro-4-fluorobenzene3-Bromo-4-fluorochlorobenzene

ACTIAS40174G100EA 0 CHF
ACTIAS40174G100
2-Bromo-4-chloro-1-fluorobenzene
2-Bromo-4-chloro-1-fluorobenzene
Formula: C₆H₃BrClF
MW: 209.44 g/mol
MDL Number: MFCD00672934
CAS Number: 1996-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR