5-Bromo-2-chloro-3-fluoropyridine

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS51146G5EA 0 CHF
ACTIAS51146G25 ACTIAS51146G100 ACTIAS51146G5
5-Bromo-2-chloro-3-fluoropyridine
5-Bromo-2-chloro-3-fluoropyridine
Formula: Câ‚…Hâ‚‚BrClFN
MW: 210.43 g/mol
MDL Number: MFCD03411574
CAS Number: 831203-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR