3-Bromo-6-chloro-2-methylpyridine

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 5-Bromo-2-chloro-6-methylpyridine5-Bromo-2-chloro-6-picoline3-Bromo-6-chloro-2-picoline

ACTIAS40327G5EA 0 CHF
ACTIAS40327G5
3-Bromo-6-chloro-2-methylpyridine
3-Bromo-6-chloro-2-methylpyridine
Formula: C₆H₅BrClN
MW: 206.47 g/mol
MDL Number: MFCD07375117
CAS Number: 132606-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR