3-Bromo-2-chloro-5-methylpyridine

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 3-Bromo-2-chloro-5-picoline

ACTIAS40314G5EA 0 CHF
ACTIAS40314G25 ACTIAS40314G5
3-Bromo-2-chloro-5-methylpyridine
3-Bromo-2-chloro-5-methylpyridine
Formula: C₆H₅BrClN
MDL Number: MFCD01830664
CAS Number: 17282-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR