3-Bromosalicylaldehyde
Supplier: ACTIVATE SCIENTIFIC GMBH
Synonyms:
3-Bromo-2-hydroxybenzaldehyde
ACTIAS30567G1EA
0
CHF
ACTIAS30567G1
ACTIAS30567G5
3-Bromosalicylaldehyde
3-Bromosalicylaldehyde
Formula:
C₇H₅BrO₂ MW: 201.02 g/mol |
MDL Number:
MFCD00016587 CAS Number: 1829-34-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...