2,5-Dibromo-4-picoline

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 2,5-Dibromo-4-methylpyridine

ACTIAS4033G5EA 30 CHF
ACTIAS4033G5
2,5-Dibromo-4-picoline
2,5-Dibromo-4-picoline
Formula: C₆H₅Br₂N
MW: 250.92 g/mol
MDL Number: MFCD00234955
CAS Number: 3430-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR