3,6-Dibromocarbazole

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 3,6-Dibromo-9H-carbazole

ACTIAS40298G5EA 0 CHF
ACTIAS40298G10 ACTIAS40298G5
3,6-Dibromocarbazole
3,6-Dibromocarbazole
Formula: C₁₂H₇Br₂N
MW: 325 g/mol
MDL Number: MFCD00004961
CAS Number: 6825-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR