2-Bromfuran
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS24335G5EA
135
CHF
ACTIAS24335G100
ACTIAS24335G5
ACTIAS24335G25
2-Bromfuran
2-Bromofuran
Formel:
C₄H₃BrO |
MDL Number:
MFCD00216609 CAS-Nummer: 584-12-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...