Samarium trinitrate hexahydrate

Danger

Synonyms: Samarium (III) nitrate hexahydrateSamarium(III) nitrate hydrate

ACTIAS23473G100EA 0 CHF
ACTIAS23473G100
Samarium trinitrate hexahydrate
Samarium trinitrate hexahydrate
Formula: N₃O₉Sm·6H₂O
MW: 444.47 g/mol
Melting Pt: 78 °C
Density: 2.37 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149857
CAS Number: 13759-83-6
EINECS: 233-798-6
UN: 1477
ADR: 5.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR