Sulphuric acid 1 mol/l (2 N) in aqueous solution

Warning

PRLS89290.460EA 0 CHF
PRLS89290.460
Sulphuric acid 1 mol/l (2 N) in aqueous solution
Sulphuric acid
Formula: H₂SO₄
MW: 98.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR