(±)-1,2,4-Butantriol
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS20750G100EA
0
CHF
ACTIAS20750G500
ACTIAS20750G100
(±)-1,2,4-Butantriol
(±)-1,2,4-Butantriol
Formel:
C₄H₁₀O₃ MW: 106,12 g/mol Siedepunkt: 304 °C (1013 hPa) Melting Pt: –20 °C Dichte: 1,21 g/cm³ (0 °C) Lager Temperatur: Raumtemperatur |
CAS-Nummer:
3068-00-6 EINECS: 221-323-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...