4-Amino-3-brom-2-chlorpyridin
Lieferant: ACTIVATE SCIENTIFIC GMBH
Synonyms:
3-Brom-2-chlor-4-pyridylamin
ACTIAS16236G1EA
0
CHF
ACTIAS16236G1
ACTIAS16236G5
4-Amino-3-brom-2-chlorpyridin
4-Amino-3-bromo-2-chloropyridine
Formel:
C₅H₄BrClN₂ MW: 207,46 g/mol |
MDL Number:
MFCD09263478 CAS-Nummer: 215364-85-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...