3-Amino-5-bromo-2-chloropyridine

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 5-Bromo-2-chloro-3-pyridylamine

ACTIAS40305G5EA 0 CHF
ACTIAS40305G5 ACTIAS40305G25
3-Amino-5-bromo-2-chloropyridine
3-Amino-5-bromo-2-chloropyridine
Formula: Câ‚…Hâ‚„BrClNâ‚‚
MW: 207.46 g/mol
MDL Number: MFCD02682092
CAS Number: 588729-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR