3-Amino-2-bromo-5-chloropyridine

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 2-Bromo-5-chloro-3-pyridylamine

ACTIAS4070G5EA 0 CHF
ACTIAS4070G5 ACTIAS4070G25
3-Amino-2-bromo-5-chloropyridine
3-Amino-2-bromo-5-chloropyridine
Formula: Câ‚…Hâ‚„BrClNâ‚‚
MDL Number: MFCD07368976
CAS Number: 90902-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR