5-Chloro-2-fluorophenol

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS31196G5EA 0 CHF
ACTIAS31196G1 ACTIAS31196G5
5-Chloro-2-fluorophenol
5-Chloro-2-fluorophenol
Formula: C₆H₄ClFO
CAS Number: 186589-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR