4-Brom-3-chlorphenol
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS34919G5EA
0
CHF
ACTIAS34919G5
ACTIAS34919G1
ACTIAS34919G10
4-Brom-3-chlorphenol
4-Brom-3-chlorphenol
Formel:
C₆H₄BrClO MW: 207,45 g/mol |
MDL Number:
MFCD00070744 CAS-Nummer: 13631-21-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...