4-Hydroxyphthalic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS40454G5EA 0 CHF
ACTIAS40454G5
4-Hydroxyphthalic acid
4-Hydroxyphthalic acid
Formula: C₈H₆O₅
MW: 182.13 g/mol
Boiling Pt: 386 °C (1013 hPa)
Melting Pt: 204 °C
MDL Number: MFCD00013984
CAS Number: 610-35-5
EINECS: 210-221-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR