4-Bromphthalsäure
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS32923G1EA
0
CHF
ACTIAS32923G1
ACTIAS32923G5
4-Bromphthalsäure
4-Bromphthalsäure
Formel:
C₈H₅BrO₄ MW: 245,03 g/mol |
MDL Number:
MFCD00043538 CAS-Nummer: 6968-28-1 EINECS: 230-183-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...