4-Fluor-3-(trifluormethyl)benzonitril

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS22439G25EA 0 CHF
ACTIAS22439G25 ACTIAS22439G100
4-Fluor-3-(trifluormethyl)benzonitril
4-Fluor-3-(trifluormethyl)benzonitril
Formel: C₈H₃F₄N
MW: 189,11 g/mol
Siedepunkt: 193…195 °C (1013 hPa)
Melting Pt: 67…68 °C
MDL Number: MFCD00061284
CAS-Nummer: 67515-59-7
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR