5-Bromo-2-thiophenesulphonyl chloride

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 5-Bromothiophene-2-sulphonyl chloride

ACTIAS35017G5EA 56.23 CHF
ACTIAS35017G5 ACTIAS35017G10 ACTIAS35017G1
5-Bromo-2-thiophenesulphonyl chloride
5-Bromo-2-thiophenesulphonyl chloride
Formula: C₄H₂BrClO₂S₂
MW: 261.55 g/mol
MDL Number: MFCD00084909
CAS Number: 55854-46-1
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR