Methyl-2-bromo-5-nitrobenzoate

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: Methyl 2-bromo-5-nitrobenzoate2-Bromobenzoic acid methyl ester

ACTIAS34458G100EA 0 CHF
ACTIAS34458G100 ACTIAS34458G10
Methyl-2-bromo-5-nitrobenzoate
Methyl 2-bromo-5-nitrobenzoate
Formula: C₈H₆BrNO₄
MW: 260.04 g/mol
Melting Pt: 82 °C
MDL Number: MFCD00010867
CAS Number: 6942-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR