4-Pyridineboronic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 4-Pyridinylboronic acidPyridine-4-boronic acidPyridin-4-ylboronic acid

ACTIAS50991G5EA 0 CHF
ACTIAS50991G1 ACTIAS50991G25 ACTIAS50991G5
4-Pyridineboronic acid
4-Pyridineboronic acid
Formula: C₅H₆BNO₂
MW: 122.92 g/mol (anhy)
MDL Number: MFCD01074545
CAS Number: 1692-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR