2-Fluoro-5-iodobenzonitrile

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS35080G5EA 0 CHF
ACTIAS35080G10 ACTIAS35080G5 ACTIAS35080G1
2-Fluoro-5-iodobenzonitrile
2-Fluoro-5-iodobenzonitrile
Formula: C₇H₃FIN
MW: 247.01 g/mol
MDL Number: MFCD03094225
CAS Number: 351003-36-6
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR