2-Chloro-5-pyridineboronic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: (6-Chloropyridin-3-yl)boronic acid6-Chloropyridine-3-boronic acid6-Chloro-3-pyridinylboronic acid2-Chloro-5-pyridinylboronic acid2-Chloropyridine-5-boronic acid

ACTIAS51139G5EA 0 CHF
ACTIAS51139G25 ACTIAS51139G5 ACTIAS51139G1
2-Chloro-5-pyridineboronic acid
2-Chloro-5-pyridineboronic acid
Formula: Câ‚…Hâ‚…BClNOâ‚‚
MW: 157.36 g/mol
MDL Number: MFCD03094998
CAS Number: 444120-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR