2-Fluoro-6-hydroxybenzonitrile
Supplier: ACTIVATE SCIENTIFIC GMBH
ACTIAS31471G1EA
0
CHF
ACTIAS31471G1
ACTIAS31471G5
2-Fluoro-6-hydroxybenzonitrile
2-Fluoro-6-hydroxybenzonitrile
Formula:
C₇H₄FNO MW: 137.11 g/mol |
MDL Number:
MFCD03428592 CAS Number: 140675-43-0 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...