2-Fluoro-5-nitrobenzonitrile

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS20089G25EA 0 CHF
ACTIAS20089G25 ACTIAS20089G100
2-Fluoro-5-nitrobenzonitrile
2-Fluoro-5-nitrobenzonitrile
Formula: C₇H₃FN₂O₂
MW: 166.11 g/mol
Melting Pt: 79 °C
MDL Number: MFCD00042299
CAS Number: 17417-09-3
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR