2-Fluoro-4-nitrobenzonitrile

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS36146G1EA 0 CHF
ACTIAS36146MG250 ACTIAS36146G1
2-Fluoro-4-nitrobenzonitrile
2-Fluoro-4-nitrobenzonitrile
Formula: C₇H₃FN₂O₂
CAS Number: 34667-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR