2-Amino-3-chlorobenzonitrile

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS40150G5EA 0 CHF
ACTIAS40150G5
2-Amino-3-chlorobenzonitrile
2-Amino-3-chlorobenzonitrile
Formula: C₇H₅ClN₂
MDL Number: MFCD03425881
CAS Number: 53312-77-9
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR