p-Tolylacetonitril
Lieferant: ACTIVATE SCIENTIFIC GMBH
Synonyms:
(4-Methylphenyl)acetonitril
, 4-Methylbenzylcyanid
ACTIAS21708G100EA
173
CHF
ACTIAS21708G100
p-Tolylacetonitril
p-Tolylacetonitril
Formel:
C₉H₉N MW: 131,18 g/mol Siedepunkt: 243 °C (1013 hPa) Melting Pt: 18 °C Dichte: 0,99 g/cm³ (20 °C) |
MDL Number:
MFCD00001922 CAS-Nummer: 2947-61-7 EINECS: 220-963-2 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...