(±)-Nipecotic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: Nipecotic acidPiperidine-3-carboxylic acid(±)-3-Piperidinecarboxylic acidH-DL-Nip-OH

ACTIAS20164G100EA 99 CHF
ACTIAS20164G100
(±)-Nipecotic acid
(±)-Nipecotic acid
Formula: C₆H₁₁NO₂
MW: 129.16 g/mol
MDL Number: MFCD00005992
CAS Number: 498-95-3
EINECS: 207-873-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR