5-Hydroxynicotinic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 5-Hydroxy-3-pyridinecarboxylic acid5-Hydroxypyridine-3-carboxylic acid

ACTIAS7147G100EA 0 CHF
ACTIAS7147G100 ACTIAS7147G5 ACTIAS7147G25
5-Hydroxynicotinic acid
5-Hydroxynicotinic acid
Formula: C₆H₅NO₃
MW: 139.11 g/mol
MDL Number: MFCD00129117
CAS Number: 27828-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR