5-Chloronicotinic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS1614G5EA 0 CHF
ACTIAS1614G100 ACTIAS1614G5
5-Chloronicotinic acid
5-Chloronicotinic acid
Formula: C₆H₄ClNO₂
MDL Number: MFCD00128901
CAS Number: 22620-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR