4-Chloro-2-fluorophenylboronic acid
Supplier: ACTIVATE SCIENTIFIC GMBH
ACTIAS32277G5EA
0
CHF
ACTIAS32277G10
ACTIAS32277G1
ACTIAS32277G5
4-Chloro-2-fluorophenylboronic acid
4-Chloro-2-fluorophenylboronic acid
Formula:
C₆H₅BClFO₂ MW: 174.37 g/mol |
MDL Number:
MFCD02684293 CAS Number: 160591-91-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...