4-Aminonicotinic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 4-Amino-3-pyridinecarboxylic acid

ACTIAS7152G100EA 0 CHF
ACTIAS7152G5 ACTIAS7152G100
4-Aminonicotinic acid
4-Aminonicotinic acid
Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
MDL Number: MFCD00234183
CAS Number: 7418-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR