3-Acetylphenylboronic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: (3-Acetylphenyl)metaboric acid3-Acetylbenzeneboronic acidDihydroxy(3-acetylphenyl)borane

ACTIAS32786G5EA 0 CHF
ACTIAS32786G10 ACTIAS32786G5
3-Acetylphenylboronic acid
3-Acetylphenylboronic acid
Formula: C₈H₉BO₃
MW: 163.97 g/mol
MDL Number: MFCD01074678
CAS Number: 204841-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR