2-Brom-5-fluorbenzylalkohol
Lieferant: ACTIVATE SCIENTIFIC GMBH
Synonyms:
(2-Brom-5-fluorphenyl)methanol
ACTIAS40008G100EA
0
CHF
ACTIAS40008G100
2-Brom-5-fluorbenzylalkohol
2-Brom-5-fluorbenzylalkohol
Formel:
C₇H₆BrFO MW: 205,03 g/mol |
MDL Number:
MFCD00142876 CAS-Nummer: 202865-66-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...