1,4-Phenyldiboronic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 1,4-Benzenediboronic acidp-Phenylenebisboronic acid1,4-Phenyldiboric acid

ACTIAS33821G5EA 0 CHF
ACTIAS33821G10 ACTIAS33821G1 ACTIAS33821G5
1,4-Phenyldiboronic acid
1,4-Phenyldiboronic acid
Formula: C₆H₈B₂O₄
MW: 165.75 g/mol
MDL Number: MFCD00236018
CAS Number: 4612-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR